Orders LinkContactUsLinkComponent
Non-return valve
Non-return valve
6 - 9 mm ID
Catalog # 229-3311
Supplier:   Brand
Non-return valve
Catalog # 229-3311
Supplier:  Brand
Non-return valve
Non-return valve
6 - 9 mm ID
Price
4,40 €
Log in for availability
1 * 1 ST
Product Details & Documents
HDPE, transparent. Prevents backflow. With FKM valve disc not suitable for pressure applications.
  • Good chemical resistance
  • Good thermal resistance
  • With tapered ends to ensure that the tube is held firmly
Specifications
  • Tubing size:
    6 - 9 mm ID