Orders LinkContactUsLinkComponent
Spherical ground joints
Spherical ground joints
Supplier:   Lenz Laborglas
Spherical ground joints
Supplier:  Lenz Laborglas
Certificates
Spherical ground joints
Spherical ground joints
10 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, clear.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
Spherical cup and ball joint with inch markings. Fused edges. Excellent chemical resistance. High temperature resistance. Minimal thermal expansion giving relatively high resistance to temperature changes.
DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641
Certifications: DIN 12244 (201-4215 and 201-0128 in addition to DIN), ISO 641.
Documents
Product Family Options
Socket members
Product Information
  • Total length
    Ball
    Socket
    Availability
    Price
  • 100 mm
    -
    S 13/5
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,10 €
    11,10 €
    1 * 1 ST
       11,10 €
  • 120 mm
    -
    S 19
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 120 mm
    -
    S 29
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
  • 120 mm
    -
    S 35
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 120 mm
    -
    S 41
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,50 €
    26,50 €
    1 * 1 ST
       26,50 €
Ball members
Product Information
  • Total length
    Ball
    Socket
    Availability
    Price
  • 120 mm
    S 13/5
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,20 €
    10,20 €
    1 * 1 ST
       10,20 €
  • 120 mm
    S 19
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,60 €
    10,60 €
    1 * 1 ST
       10,60 €
  • 120 mm
    S 29
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,00 €
    14,00 €
    1 * 1 ST
       14,00 €
  • 120 mm
    S 35
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,50 €
    15,50 €
    1 * 1 ST
       15,50 €
  • 120 mm
    S 41
    -
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,50 €
    27,50 €
    1 * 1 ST
       27,50 €