Caps, BACTI-CAPS for reagent and culture tubes
Supplier: MHP Industries
Certificates
About this item
PP, coloured.
- Seal the tubes completely yet allow exchange of controlled gases
- Excellent chemical resistance, autoclavable
- Available colour-coded for tubes of different sizes
For smooth-edged glass or plastic reagent and culture tubes.
Ideal for cell culture. The alternative to conventional cotton and pulp stoppers.
Frequently Bought Together