Orders LinkContactUsLinkComponent
Closures for test tubes and flasks
Closures for test tubes and flasks
Supplier:   JAECE INDUSTRIES
Closures for test tubes and flasks
Supplier:  JAECE INDUSTRIES
Certificates
Closures for test tubes and flasks
Closures for test tubes and flasks
7 Options Available Below
Product Details & Documents
Identy-plug.
Plastic/foam plugs provide a snug fit, yet permit air flow; springs back to original shape after repeated use; autoclavable and chemically resistant.
Cotton cheesecloth.
100% pure cotton, very absorbent and low linting; 28×24 thread count.
Bleached cotton balls.
Available large or medium.
Product Family Options
Identi-plug
Product Information
  • Availability
    Price
  • JAECL800-A
    Identi-plug foam plugs, Ø 6 - 13 mm
    Identi-plug foam plugs, Ø 6 - 13 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 174,00 €
    174,00 €
    1 * 1.000 ST
       174,00 €
  • JAECL800-B
    Identi-plug foam plugs, Ø 14 - 20 mm
    Identi-plug foam plugs, Ø 14 - 20 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 214,00 €
    214,00 €
    1 * 1.000 ST
       214,00 €
  • 734-1262
    Identi-plug foam plugs, Ø 21 - 26 mm
    Identi-plug foam plugs, Ø 21 - 26 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 188,00 €
    188,00 €
    1 * 400 ST
       188,00 €
  • JAECL800-D
    Identi-plug foam plugs, Ø 35 - 45 mm
    Identi-plug foam plugs, Ø 35 - 45 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 104,00 €
    104,00 €
    1 * 100 ST
       104,00 €
Cotton cheesecloth
Product Information
  • Availability
    Price
  • 734-1264
    Cotton cheesecloth, grade 50, 24 - 28 mm
    Cotton cheesecloth, grade 50, 24 - 28 mm
    Log in for availability
    Log in for availability
    1 * 700 ST
    1 * 700 ST - 1 158,48 €
    1 158,48 €
    1 * 700 ST
       1 158,48 €
This product has been blocked by your organization. Please contact your purchasing department for more information
Bleached cotton balls
Product Information
  • Availability
    Price
  • 734-1266
    Cotton balls, medium, non sterile
    Cotton balls, medium, non sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 44,81 €
    44,81 €
    1 * 2.000 ST
       44,81 €
  • 734-1265
    Cotton balls, large, non sterile
    Cotton balls, large, non sterile
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 89,61 €
    89,61 €
    1 * 2.000 ST
       89,61 €
This product has been blocked by your organization. Please contact your purchasing department for more information
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.