Orders LinkContactUsLinkComponent
VWR®, Closures
VWR®, Closures
VWR®, Closures
Supplier:  VWR Collection
Certificates
VWR®, Closures
2 Options Available Below
Product Details & Documents
These closures are suitable for test and culture tubes.
  • Flexible, high mechanical resistance
Closures are made from LDPE.
Product Family Options
Product Information
  • Material
    For tubes
    Colour
    Cap Size
    Availability
    Price
  • LDPE
    212-1809/212-1837
    Natural
    10 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 92,30 €
    92,30 €
    1 * 3.000 ST
       92,30 €
  • LDPE
    212-1834
    Natural
    11 mm
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 66,60 €
    66,60 €
    1 * 2.500 ST
       66,60 €
Complementary Products
Product Information
  • Catalog #
    Capacity
    Length
    Material
    Ø int.
    Ext.Ø
    Availability
    Price
  • 212-1834
    5 ml
    74,5 mm
    PS
    12,8 mm
    13,0 mm
    Log in for availability
    1 * 4.000 ST - 147,00 €
  • 212-1837
    3,5 ml
    PS
    11,8 mm
    01
    Log in for availability
    1 * 5.000 ST - 166,00 €
  • 212-1809
    5 ml
    74,5 mm
    PS
    11,0 mm
    13,0 mm
    Log in for availability
    1 * 2.000 ST - 105,00 €