Orders LinkContactUsLinkComponent
Test and sample tubes
Test and sample tubes
Test and sample tubes
Supplier:  DELTALAB
Certificates
Test and sample tubes
Test and sample tubes
9 Options Available Below
Product Details & Documents
Clear polystyrene (PS).
  • Temperature resistance from −10 up to +70 °C (for short periods up to +80 °C)
  • Can be centrifuged up to 7500×g
  • Tubes 212-3304 graduated to 2,5/5,0 and 10 ml; tubes 212-3302 graduated to 1,25 and 5,0 ml
Precision moulded sample tubes for a wide variety of applications. With round base. Non sterile.
Documents
Product Family Options
Product Information
  • Capacity
    Length
    Ext.Ø
    Ø int.
    Availability
    Price
  • 3 ml
    54,4 mm
    11,5 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 102,00 €
    102,00 €
    1 * 2.000 ST
       102,00 €
  • 4 ml
    69,4 mm
    10,8 mm
    9,1 mm
    Log in for availability
    Log in for availability
    1 * 3.000 ST
    1 * 3.000 ST - 180,00 €
    180,00 €
    1 * 3.000 ST
       180,00 €
  • 5 ml
    74,5 mm
    11,8 mm
    10,3 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 53,60 €
    53,60 €
    1 * 1.000 ST
       53,60 €
  • 6 ml
    87,6 mm
    12 mm
    10,4 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 67,90 €
    67,90 €
    1 * 1.000 ST
       67,90 €
  • 5 ml
    74,6 mm
    13 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 65,40 €
    65,40 €
    1 * 1.000 ST
       65,40 €
  • 7 ml
    99,6 mm
    12,9 mm
    11 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 10 ml
    96,5 mm
    15,6 mm
    13,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 51,90 €
    51,90 €
    1 * 500 ST
       51,90 €
  • 11 ml
    99,8 mm
    16 mm
    13,8 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 62,30 €
    62,30 €
    1 * 500 ST
       62,30 €
  • 17 ml
    151 mm
    16,3 mm
    14,6 mm
    Log in for availability
    Log in for availability
    1 * 850 ST
    1 * 850 ST - 220,00 €
    220,00 €
    1 * 850 ST
       220,00 €
Closures
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Red
    Cap for 11 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 99,50 €
    99,50 €
    1 * 2.000 ST
       99,50 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 15 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 116,00 €
    116,00 €
    1 * 2.500 ST
       116,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Green
    Cap for 12 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 84,20 €
    84,20 €
    1 * 2.000 ST
       84,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Green
    Cap for 16 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 103,00 €
    103,00 €
    1 * 2.500 ST
       103,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 16 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 102,00 €
    102,00 €
    1 * 2.500 ST
       102,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Blue
    Cap for 12 and 13 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 58,20 €
    58,20 €
    1 * 2.000 ST
       58,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Green
    Cap for 12 and 13 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 58,20 €
    58,20 €
    1 * 2.000 ST
       58,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 12 and 13 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 58,20 €
    58,20 €
    1 * 2.000 ST
       58,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Red
    Cap for 15 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 102,00 €
    102,00 €
    1 * 2.500 ST
       102,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 15 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 102,00 €
    102,00 €
    1 * 2.500 ST
       102,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Mauve
    Cap for 11 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 99,50 €
    99,50 €
    1 * 2.000 ST
       99,50 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 12 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 84,20 €
    84,20 €
    1 * 2.000 ST
       84,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Green
    Cap for 11 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 99,50 €
    99,50 €
    1 * 2.000 ST
       99,50 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Blue
    Cap for 12 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 84,20 €
    84,20 €
    1 * 2.000 ST
       84,20 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Green
    Cap for 15 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.500 ST
    1 * 2.500 ST - 102,00 €
    102,00 €
    1 * 2.500 ST
       102,00 €
Product Information
  • Catalog #
    Colour
    Description
    Availability
    Price
  • Natural
    Cap for 11 mm tubes
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 99,50 €
    99,50 €
    1 * 2.000 ST
       99,50 €
Product Information
  • Catalog #
    Description
    Availability
    Price
  • Screw cap
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 55,30 €
    55,30 €
    1 * 1.000 ST
       55,30 €