Test tubes with rim, DURAN®
Supplier: DWK Life Sciences
Certificates
About this item
FIOLAX®, neutral glass.
- Excellent chemical resistance
- Relatively resistant against rapid temperature changes and local heating
Thin walled test tubes for a wide variety of applications.
Frequently Bought Together