Cap strips, flat top
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Made from polypropylene (PP).
- Flat surface of each cap facilitates sample labelling
- Large labelled end tabs for strip orientation and removal (A-H for 8 strip, 1 to 12 for 12 strip)
- Certified free from DNase, RNase and Human DNA
Suitable for use with most of the 96-well PCR plates, as well as standard and low profile tube strips.
Colour: Natural
5 Options Available Below
Product Details & Documents
Made from polypropylene (PP).
- Flat surface of each cap facilitates sample labelling
- Large labelled end tabs for strip orientation and removal (A-H for 8 strip, 1 to 12 for 12 strip)
- Certified free from DNase, RNase and Human DNA
Suitable for use with most of the 96-well PCR plates, as well as standard and low profile tube strips.
Colour: Natural