ContactUsLinkComponent
Microcentrifuge tubes, low surface tension, Cliklok™
Microcentrifuge tubes, low surface tension, Cliklok™
Catalog # SIMPT330-7LST
Supplier:  Simport Scientific
undefined
Microcentrifuge tubes, low surface tension, Cliklok™
Catalog # SIMPT330-7LST
Supplier:  Simport Scientific
Specifications
  • Type:
    with attached Cliklok™ lid, , low surface tension
  • Version:
    graduated, conical bottom, non sterile
  • Capacity:
    1,5 ml
  • Colour:
    Colourless
  • Pk:
    500
  • RCF max.:
    20000×g
Specifications
About this item
PP, graduated.
  • One-piece construction with snug fitting attached cap and reinforced tube bottom for added protection against leakage
  • No lubricant (e.g. silicone) needed, thereby eliminating the danger of sample contamination
  • Designed to withstand the stress of high speed centrifugation up to 20000×g
Low adhesion surface for optimum sample yield. Cliklok™ seal is especially designed to reduce popping where freezing, boiling, storing, or shipping is required.
Packaging: Packaged in tamper-proof resealable safety-lock bags of 500 tubes. 10 bags per case.