Order Entry
Belgium
Orders LinkContactUsLinkComponent
Coloured cap inserts

Coloured cap inserts

Supplier:   Brand
Coloured cap inserts
Supplier:  Brand
Certificates
Coloured cap inserts
Coloured cap inserts

5 Options Available Below

Product Details & Documents

PP.

  • For quick identification
  • For microtubes 211-3201 to 211-3205 and 211-3211 to 211-3215
  • Applications: −196 to +121 °C
Product Family Options

Product Information

  • Colour
    Availability
    Price
  • Blue
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 21,80 €
    21,80 €
    1 * 500 ST
       21,80 €

  • Green
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 21,80 €
    21,80 €
    1 * 500 ST
       21,80 €

  • Red
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 21,80 €
    21,80 €
    1 * 500 ST
       21,80 €

  • White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 21,80 €
    21,80 €
    1 * 500 ST
       21,80 €

  • Yellow
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 21,80 €
    21,80 €
    1 * 500 ST
       21,80 €