Micro Dialysis System, Tube-O-Dialyzer™
Supplier: G-Biosciences
Certificates
About this item
'NO-LOSS' dialyser.
The sample container itself serves as the Tube-O-DIALYZER™, whereby the cap is fitted with a dialysis membrane made from a neutral non binding membrane. Samples are recovered through brief centrifugation. For storing the sample, the dialysis cap can be replaced with a normal cap.
Developed for the dialysis of small but valuable samples. The complete procedure is carried out in special tubes avoiding the need to transfer samples and therefore also avoiding the loss of samples.