Orders LinkContactUsLinkComponent
Culture tubes, Rimless
Culture tubes, Rimless
Supplier:   Brand
Culture tubes, Rimless
Supplier:  Brand
Certificates
Culture tubes, Rimless
Culture tubes, Rimless
6 Options Available Below
Product Details & Documents
Made of AR-Glas®.
  • Autoclavable (at 121 °C/2 bar)
  • Wall thickness 0,6/0,7 mm
Certifications: DIN EN 285
Product Family Options
Product Information
  • Wall thickness
    Height
    Ø
    RCF max.
    Availability
    Price
  • 0,6 mm
    75 mm
    10 mm
    3000×g
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 27,10 €
    27,10 €
    1 * 250 ST
       27,10 €
  • 0,6 mm
    75 mm
    12 mm
    3000×g
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 34,20 €
    34,20 €
    1 * 250 ST
       34,20 €
  • 0,6 mm
    100 mm
    12 mm
    3000×g
    Log in for availability
    Log in for availability
    1 * 144 ST
    1 * 144 ST - 19,90 €
    19,90 €
    1 * 144 ST
       19,90 €
  • 0,7 mm
    160 mm
    16 mm
    1500×g
    Log in for availability
    Log in for availability
    1 * 344 ST
    1 * 344 ST - 76,30 €
    76,30 €
    1 * 344 ST
       76,30 €
  • 0,7 mm
    100 mm
    16 mm
    2600×g
    Log in for availability
    Log in for availability
    1 * 78 ST
    1 * 78 ST - 15,20 €
    15,20 €
    1 * 78 ST
       15,20 €
  • 0,7 mm
    180 mm
    18 mm
    900×g
    Log in for availability
    Log in for availability
    1 * 121 ST
    1 * 121 ST - 44,80 €
    44,80 €
    1 * 121 ST
       44,80 €