Orders LinkContactUsLinkComponent
Centrifuge tubes
Centrifuge tubes
Centrifuge tubes
Supplier:  Greiner Bio-One
Certificates
Centrifuge tubes
Centrifuge tubes
4 Options Available Below
Product Details & Documents
Transparent PS or PP, 2-position ventilation stopper, round bottom, sterile.
  • PS tubes without graduations, good thermal, mechanical and chemical resistance, centrifugation RFC 5000×g (fix-angle rotor) or 4800×g (swinging-bucket rotor)
  • PP tubes with graduations; high thermal, mechanical and chemical resistance, centrifugation RCF 34000×g (fix-angle rotor) or 5800×g (swinging-bucket rotor)
  • The ventilation stopper has two functions: when pushed in gently, it provides uniform ventilation of the tube contents; when pushed in firmly, it provides a tight seal
  • Free from DNase/RNase and human DNA, non-pyrogenic
Ordering information: Available in packs of 25 or wrapped individually.
Documents
Product Family Options
PS tubes, non graduated
Product Information
  • Capacity
    Ø×L
    Closure type
    Cap colour
    Type
    Version
    Packed
    Availability
    Price
  • 4,5 ml
    12,4×75 mm
    Vented cap
    Transparent
    Round bottom
    sterile
    Pack of 25
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 382,00 €
    382,00 €
    1 * 2.000 ST
       382,00 €
  • 14 ml
    18×95 mm
    Vented cap
    Transparent
    Round bottom
    sterile
    Pack of 25
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 343,00 €
    343,00 €
    1 * 1.000 ST
       343,00 €
PP tubes, graduated
Product Information
  • Capacity
    Ø×L
    Closure type
    Cap colour
    Type
    Version
    Packed
    Availability
    Price
  • 14 ml
    18×95 mm
    Vented cap
    Natural
    Round bottom, with writing area
    sterile
    Packs of 25
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 302,00 €
    302,00 €
    1 * 1.000 ST
       302,00 €
  • 5 ml
    12×75 mm
    Vented cap
    Natural
    Round bottom
    Sterile
    Packs of 25
    Log in for availability
    Log in for availability
    1 * 2.000 ST
    1 * 2.000 ST - 577,00 €
    577,00 €
    1 * 2.000 ST
       577,00 €