Orders LinkContactUsLinkComponent
Transfer capillaries
Transfer capillaries
Supplier:   HIRSCHMANN
Transfer capillaries
Supplier:  HIRSCHMANN
Certificates
Transfer capillaries
Transfer capillaries
36 Options Available Below
Product Details & Documents
Clear glass, both end open.
Product Family Options
Product Information
  • Length
    Ext.Ø
    Ø int.
    Availability
    Price
  • 70 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 91,40 €
    91,40 €
    1 * 1.000 ST
       91,40 €
  • 75 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,10 €
    92,10 €
    1 * 1.000 ST
       92,10 €
  • 80 mm
    1,00 mm
    0,80
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,40 €
    92,40 €
    1 * 1.000 ST
       92,40 €
  • 90 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,90 €
    92,90 €
    1 * 1.000 ST
       92,90 €
  • 100 mm
    1,00 mm
    0,80
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 107,00 €
    107,00 €
    1 * 1.000 ST
       107,00 €
  • 120 mm
    1,00 mm
    0,80
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 109,00 €
    109,00 €
    1 * 1.000 ST
       109,00 €
  • 150 mm
    1,00 mm
    0,80 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 118,00 €
    118,00 €
    1 * 1.000 ST
       118,00 €
  • 60 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 63,70 €
    63,70 €
    1 * 1.000 ST
       63,70 €
  • 70 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 63,70 €
    63,70 €
    1 * 1.000 ST
       63,70 €
  • 75 mm
    1,35 mm
    0,95
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 64,20 €
    64,20 €
    1 * 1.000 ST
       64,20 €
  • 80 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 64,60 €
    64,60 €
    1 * 1.000 ST
       64,60 €
  • 90 mm
    1,35
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 67,20 €
    67,20 €
    1 * 1.000 ST
       67,20 €
  • 100 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 74,70 €
    74,70 €
    1 * 1.000 ST
       74,70 €
  • 120 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 87,50 €
    87,50 €
    1 * 1.000 ST
       87,50 €
  • 150 mm
    1,35 mm
    0,95 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 93,30 €
    93,30 €
    1 * 1.000 ST
       93,30 €
  • 70 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 65,90 €
    65,90 €
    1 * 1.000 ST
       65,90 €
  • 75 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 66,40 €
    66,40 €
    1 * 1.000 ST
       66,40 €
  • 80 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 67,50 €
    67,50 €
    1 * 1.000 ST
       67,50 €
  • 90 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 68,40 €
    68,40 €
    1 * 1.000 ST
       68,40 €
  • 100 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 69,00 €
    69,00 €
    1 * 1.000 ST
       69,00 €
  • 120 mm
    1,55 mm
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 88,80 €
    88,80 €
    1 * 1.000 ST
       88,80 €
  • 70 mm
    1,75
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 72,70 €
    72,70 €
    1 * 1.000 ST
       72,70 €
  • 150
    1,55
    1,15 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 91,30 €
    91,30 €
    1 * 1.000 ST
       91,30 €
  • 75 mm
    1,75 mm
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,10 €
    92,10 €
    1 * 1.000 ST
       92,10 €
  • 80 mm
    1,75 mm
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 93,40 €
    93,40 €
    1 * 1.000 ST
       93,40 €
  • 90 mm
    1,75 mm
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 100,00 €
    100,00 €
    1 * 1.000 ST
       100,00 €
  • 100 mm
    1,75 mm
    1,30
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 85,80 €
    85,80 €
    1 * 1.000 ST
       85,80 €
  • 120 mm
    1,75 mm
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 103,00 €
    103,00 €
    1 * 1.000 ST
       103,00 €
  • 150 mm
    1,75 mm
    1,30 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 104,00 €
    104,00 €
    1 * 1.000 ST
       104,00 €
  • 70 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 92,70 €
    92,70 €
    1 * 1.000 ST
       92,70 €
  • 75 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 96,40 €
    96,40 €
    1 * 1.000 ST
       96,40 €
  • 80 mm
    2,35 mm
    1,55
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 102,00 €
    102,00 €
    1 * 1.000 ST
       102,00 €
  • 90 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 108,00 €
    108,00 €
    1 * 1.000 ST
       108,00 €
  • 100 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 130,00 €
    130,00 €
    1 * 1.000 ST
       130,00 €
  • 120 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 165,00 €
    165,00 €
    1 * 1.000 ST
       165,00 €
  • 150 mm
    2,35 mm
    1,55 mm
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 185,00 €
    185,00 €
    1 * 1.000 ST
       185,00 €