Orders LinkContactUsLinkComponent
Tripod stands
Tripod stands
Tripod stands
Tripod stands
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
Product Family Options
Stainless steel 18/8
  • Int.Ø×H
  • 100×180 mm
    1 * 1 ST
    1 * 1 ST - 33,90 €
    33,90 €
    1 * 1 ST
       33,90 €
  • 120×210 mm
    1 * 1 ST
    1 * 1 ST - 36,80 €
    36,80 €
    1 * 1 ST
       36,80 €
  • 120×250 mm
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • 120×260 mm
    1 * 1 ST
    1 * 1 ST - 40,00 €
    40,00 €
    1 * 1 ST
       40,00 €
  • 140×220 mm
    1 * 1 ST
    1 * 1 ST - 42,30 €
    42,30 €
    1 * 1 ST
       42,30 €
  • 160×230 mm
    1 * 1 ST
    1 * 1 ST - 48,50 €
    48,50 €
    1 * 1 ST
       48,50 €
Zinc plated steel
  • Int.Ø×H
  • 100×180 mm
    1 * 1 ST
    1 * 1 ST - 15,60 €
    15,60 €
    1 * 1 ST
       15,60 €
  • 120×210 mm
    1 * 1 ST
    1 * 1 ST - 17,40 €
    17,40 €
    1 * 1 ST
       17,40 €
  • 120×250 mm
    1 * 1 ST
    1 * 1 ST - 22,20 €
    22,20 €
    1 * 1 ST
       22,20 €
  • 120×260 mm
    1 * 1 ST
    1 * 1 ST - 23,50 €
    23,50 €
    1 * 1 ST
       23,50 €
  • 140×220 mm
    1 * 1 ST
    1 * 1 ST - 19,70 €
    19,70 €
    1 * 1 ST
       19,70 €
  • 160×230 mm
    1 * 1 ST
    1 * 1 ST - 23,00 €
    23,00 €
    1 * 1 ST
       23,00 €