Orders LinkContactUsLinkComponent
Laboratory trays
Laboratory trays
Supplier:   KARTELL
Laboratory trays
Supplier:  KARTELL
Certificates
Laboratory trays
Laboratory trays
15 Options Available Below
Product Details & Documents
PS, white.
  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning
General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.
Documents
Product Family Options
Product Information
  • W×D×H
    Availability
    Price
  • 201×151x21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,00 €
    12,00 €
    1 * 1 ST
       12,00 €
  • 201×151×41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,10 €
    14,10 €
    1 * 1 ST
       14,10 €
  • 202×151×81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 15,30 €
    15,30 €
    1 * 1 ST
       15,30 €
  • 252×252×42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,80 €
    18,80 €
    1 * 1 ST
       18,80 €
  • 252×252×82 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • 254×254×21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 17,50 €
    17,50 €
    1 * 1 ST
       17,50 €
  • 303×151×21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,50 €
    14,50 €
    1 * 1 ST
       14,50 €
  • 303×151×42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,60 €
    16,60 €
    1 * 1 ST
       16,60 €
  • 303×151×81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • 353×252×21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,30 €
    22,30 €
    1 * 1 ST
       22,30 €
  • 353×253×41 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,40 €
    24,40 €
    1 * 1 ST
       24,40 €
  • 352×252×81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,10 €
    34,10 €
    1 * 1 ST
       34,10 €
  • 408×300×21 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,60 €
    32,60 €
    1 * 1 ST
       32,60 €
  • 409×300×42 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,80 €
    32,80 €
    1 * 1 ST
       32,80 €
  • 408×299×81 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 35,60 €
    35,60 €
    1 * 1 ST
       35,60 €