Orders LinkContactUsLinkComponent
Temperature indicating strips
Temperature indicating strips
Supplier:   TESTO
Temperature indicating strips
Supplier:  TESTO
Certificates
Temperature indicating strips
Temperature indicating strips
6 Options Available Below
Product Details & Documents
These indicators are self-adhesive, temperature proof foils with temperature sensitive elements for temperature control and regulation. They are particularly suitable for monitoring temperature in small objects.
  • Change in colour within two seconds
  • Practical booklet with 10 indicators
  • Maximum storage temperature: 25 °C; storage in a refrigerator is recommended
Product Family Options
Product Information
  • Range
    Accuracy
    Ø
    Availability
    Price
  • 620-9122
    Temperature indicator
    Temperature indicator
    60...82 °C
    ±1,5 °C
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • 620-9123
    Temperature indicator
    Temperature indicator
    88...110 °C
    ±1,5 °C
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • 620-9124
    Temperature indicator
    Temperature indicator
    116...138 °C
    ±1,5 °C
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • 620-9125
    Temperature indicator
    Temperature indicator
    143...166 °C
    ±1,5 °C
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • 620-9126
    Temperature indicator
    Temperature indicator
    171...193 °C
    ±1,0 °C ±1% mv
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
  • 620-9127
    Temperature indicator
    Temperature indicator
    199...224 °C
    ±1,0 °C ±1% mv
    15 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €