ContactUsLinkComponent
VWR®, Thermometer Armor Cases
VWR®, Thermometer Armor Cases
Catalog # 620-0900
Supplier:  VWR Collection
undefined
VWR®, Thermometer Armor Cases
Catalog # 620-0900
Supplier:  VWR Collection
Specifications
  • Description:
    Metal armor case, 254 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
This case protects your thermometers from accidental breakage during use and storage.
  • Screw cap
  • Ring for hanging
  • Metal or nickel-plated brass
  • Lower section surrounding the bulb with small openings for circulation