Order Entry
Belgium
ContactUsLinkComponent
VWR®, Bi-Metallic Dial Thermometer, Refrigerator/Freezer
VWR®, Bi-Metallic Dial Thermometer, Refrigerator/Freezer
Catalog # 620-2146
Supplier:  VWR Collection
undefined
VWR®, Bi-Metallic Dial Thermometer, Refrigerator/Freezer
Catalog # 620-2146
Supplier:  VWR Collection

Specifications

  • Ø:
    76 mm
  • Accuracy:
    1 °C
  • Pk:
    1
  • Range:
    −30…+30 °C

Specifications

About this item

This thermometer, with analogue dial, indicates freezer, refrigerator, or spoilage temperatures.

  • Stainless steel model stands sturdily on rack shelves
  • Dual scale (°F/°C)
  • Red arrow for easier reading
  • Round, black modern design