About this item
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
- Non bleeding - do not contaminate the solution to be measured
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
: Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even under changing lighting conditions. The StripScan app allows the customer to analyse the MQuant® test strips with their smartphone for fast, convenient and precise results which can be easily graphically presented and shared.
12 Options Available Below
Product Details & Documents
These test strips contain special indicator dyes that are covalently bound to the reagent papers which are then adhered to the plastic strips.
- Non bleeding - do not contaminate the solution to be measured
- Quality assurance - tests are checked and calibrated against certified buffer solutions
- Wide and narrow pH ranges available
: Analyse your pH 0 to 14 test strips (1.09535) with this reference card and the free MQuant® StripScan app. The brilliant colour-fast reference colours allow for robust measurement results even under changing lighting conditions. The StripScan app allows the customer to analyse the MQuant® test strips with their smartphone for fast, convenient and precise results which can be easily graphically presented and shared.
Specifications for Product Family
Specifications
- Catalog No:
- 1.09535.0001
- 1.09540.0001
- 1.09531.0001
- 1.09541.0001
- 1.09542.0001
- 1.09543.0001
- 1.09545.0001
- 1.09584.0001
- 1.09532.0001
- 1.09450.0010
- 1.09533.0001
- 1.09502.0001
- M-Clarity quality level:
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
- MQ100
Specifications
Product Family Options
Product Information
- pH rangepH gradationAvailabilityPrice
- 1.09535.0001Universal indicator stripsSpecifications:
More
Specifications
Cat. No.1.09535.0001M-Clarity quality levelMQ100 - 1.09540.0001Special pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09540.0001M-Clarity quality levelMQ100 - 1.09531.0001pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09531.0001M-Clarity quality levelMQ100 - 1.09541.0001Special pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09541.0001M-Clarity quality levelMQ100 - 1.09542.0001Special pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09542.0001M-Clarity quality levelMQ100 - 1.09543.0001Special pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09543.0001M-Clarity quality levelMQ100 - 1.09545.0001Special pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09545.0001M-Clarity quality levelMQ100 - 1.09584.0001pH indicator strips, mid rangeSpecifications:
More
Specifications
Cat. No.1.09584.0001M-Clarity quality levelMQ100 - 1.09532.0001pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09532.0001M-Clarity quality levelMQ100 - 1.09450.0010pH strips, individually wrapped (without colour scale)pH strips, individually wrapped (without colour scale)2,0 - 9,01Log in for availability1 * 1 EA203,00 €Specifications:
More
Specifications
Cat. No.1.09450.0010M-Clarity quality levelMQ100 - 1.09533.0001pH indicator stripsSpecifications:
More
Specifications
Cat. No.1.09533.0001M-Clarity quality levelMQ100 - 1.09502.0001pH indicator strips for turbid solutionsSpecifications:
More
Specifications
Cat. No.1.09502.0001M-Clarity quality levelMQ100