Blood typing kit and cards, Eldoncard
Supplier: ELDON CARD
Certificates
About this item
The presence or absence of agglutinates will show the blood type. The blood typing kit contains everything to type one person: 1 Eldoncard, 4 Eldonsticks, 1 alcohol swab, 2 safety lancets, 1 result.card. 2 micropipettes are supplied for 50 blood typings. Shelf-life 24 months at 5-37 °C. The foil is used to protect the cards after use.
- Portable and compact
- Blood grouping on location
- Result after just 2 minutes
- Easy to learn, easy to use
- No need for refrigeration and daily controls
- Whole blood can be used down to 5 μl
- No need for separation of red cells and blood plasma
Frequently Bought Together