Protective laboratory labelling tapes, SP Bel-Art
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Clear, transparent polyester tape. The tape dispenser is available separately.
- Remains securely affixed with no discolouration after autoclaving for 40 minutes at 125 °C or after storage for one week in liquid nitrogen at −196 °C
- Resistant to acids, alkalis and organic solvents
- Gives smudge proof labelling with outstanding adhesive properties
Length: 32,9 m
Colour: Transparent