Orders LinkContactUsLinkComponent
Cleanroom tapes
Cleanroom tapes
Supplier:   ULTRATAPE
Cleanroom tapes
Supplier:  ULTRATAPE
Certificates
Cleanroom tapes
3 Options Available Below
Product Details & Documents
These tapes are made of polyolefin with acrylic adhesive.
  • Ultra low outgassing
  • Low sulfur, low halogen and low heavy metal
  • Adhere lightly to plastics for easy removal
  • Leaves no residue and particulates
Designed specifically for use inside semiconductor and critical environment cleanrooms. It contains extremely low leachable metals, eliminating the concern of contamination.
Length: 54,8 m
Colour: White
Product Family Options
Product Information
  • Width
    Availability
    Price
  • 817-0072
    Cleanroom tape
    Cleanroom tape
    50,8 mm
    Log in for availability
    Log in for availability
    1 * 3 ROU
    1 * 3 ROU - 469,00 €
    469,00 €
    1 * 3 ROU
       469,00 €
  • 817-0073
    Cleanroom tape
    Cleanroom tape
    76,2 mm
    Log in for availability
    Log in for availability
    1 * 2 ROU
    1 * 2 ROU - 180,00 €
    180,00 €
    1 * 2 ROU
       180,00 €
  • 112-1186
    Cleanroom tape
    Cleanroom tape
    25,4 mm
    Log in for availability
    Log in for availability
    1 * 6 ROU
    1 * 6 ROU - 81,20 €
    81,20 €
    1 * 6 ROU
       81,20 €