Packaging tape, wafer box tape
These PE tapes are designed specifically for sealing wafer boxes. They are also used for sealing plastic bags and for other cleanroom uses.
- Adheres lightly to plastics, stretching easily around corners
- Low metal content
- No residues after prolonged storage
- Easy release from the roll with low static charge
Length: 33,0 m.
Different colours available.
: Supplied clean and double bagged.
Frequently Bought Together