Orders LinkContactUsLinkComponent
Sterican® Single-Use Needles
 :   B. Braun
Sterican® Single-Use Needles
 :  B. Braun
Sterican® Single-Use Needles
Sterican® Single-Use Needles
Stainless chromium-nickel steel needle with extremely smooth surface and light silicone coating, transparent Luer lock PP hub.
  • Colour coded hub, conforms to ISO 6009
  • Latex- and PVC-free
  • Tri-bevelled for minimal pain upon puncture (long bevel)
 : ISO 7864, DIN 13097
 : Individually, sterile packed. 100 needles (20 blocks of 5) per box.
Product Family Options
Sterile
  • Description
    Gauge
    Length
    Ø
    Colour code
  • Intravenous, for large volumes, intramuscular, for hydrous solutions (size 2), made in china, with Max.shelf life 60 months
    21
    40 mm
    0,80 mm
    Green
    1 * 100 ST
    1 * 100 ST - 4,80 €
    4,80 €
    1 * 100 ST
       4,80 €
  • Intravenous, intramuscular (size 12), made in china, with Max.shelf life 60 months
    22
    30 mm
    0,70 mm
    Black
    1 * 100 ST
    1 * 100 ST - 6,50 €
    6,50 €
    1 * 100 ST
       6,50 €