Orders LinkContactUsLinkComponent
Syringes, 3-piece
Syringes, 3-piece
Supplier:   TERUMO
Syringes, 3-piece
Supplier:  TERUMO
Certificates
Syringes, 3-piece
Syringes, 3-piece
11 Options Available Below
Product Details & Documents
Sterile, disposable, three-piece, latex-free.
  • Available with Luer slip tip or Luer lock threaded tip
  • Sterilised by electron beam irradiation
Certifications: Compliant with Directive 93/42/EEC Class IIa, organisation accredited by TÜV Rheinland (Germany), factory certified to ISO 9001 EN 46001, sterilisation process validated according to Standard NF EN 550.
Packaging: Individual sterile blister packs.
Product Family Options
Without needles
Product Information
  • Capacity
    Availability
    Price
  • 613-1591
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    2,5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 34,90 €
    34,90 €
    1 * 100 ST
       34,90 €
  • 613-3875
    Without needle, Luer slip tip
    Without needle, Luer slip tip
    2,5 ml
    Discontinued
  • 613-0917
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    5 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 37,90 €
    37,90 €
    1 * 100 ST
       37,90 €
  • 613-0973
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    10 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 48,40 €
    48,40 €
    1 * 100 ST
       48,40 €
  • 613-3879
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    20 ml
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 38,50 €
    38,50 €
    1 * 50 ST
       38,50 €
  • TERUSS30-L1MP
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    30 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 23,30 €
    23,30 €
    1 * 25 ST
       23,30 €
  • TERUSS30-ES1
    Without needle, Luer eccentric tip
    Without needle, Luer eccentric tip
    30 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 23,30 €
    23,30 €
    1 * 25 ST
       23,30 €
  • 613-3881
    Without needle, Luer catheter tip
    Without needle, Luer catheter tip
    50 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 27,80 €
    27,80 €
    1 * 25 ST
       27,80 €
  • 613-4902
    Without needle, Luer eccentric tip
    Without needle, Luer eccentric tip
    50 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 27,80 €
    27,80 €
    1 * 25 ST
       27,80 €
  • 613-5398
    Without needle, Luer lock tip
    Without needle, Luer lock tip
    50 ml
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 27,80 €
    27,80 €
    1 * 25 ST
       27,80 €
With needles
Product Information
  • Capacity
    Availability
    Price
  • TERUMDSS01S2516E
    With needle G25×16 mm
    With needle G25×16 mm
    1 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 22,60 €
    22,60 €
    1 * 100 ST
       22,60 €
  • TERUMDSS01S2613E
    With needle G26×13 mm
    With needle G26×13 mm
    1 ml
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 22,60 €
    22,60 €
    1 * 100 ST
       22,60 €
  • 613-4918
    With needle G21×40 mm
    With needle G21×40 mm
    5 ml
    Discontinued
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.