Orders LinkContactUsLinkComponent
Repeating dispensing system, PB600-1
Repeating dispensing system, PB600-1
Repeating dispenser, PB600-1
Catalog # 549-0233
Supplier:   HAMILTON BONADUZ
Repeating dispensing system, PB600-1
Catalog # 549-0233
Supplier:  HAMILTON BONADUZ
Repeating dispensing system, PB600-1
Repeating dispensing system, PB600-1
Repeating dispenser, PB600-1
Price
233,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
The PB600-1 works with Microliter™ or Gastight® syringes from 10 µl to 2,5 ml. Use the repeating dispenser with cemented needles (N), removable needles (RN), Luer tip (LT), or PTFE Luer lock (TLL) syringes.
  • Up to 50 times with the push of a button
  • Volumes are between 0,2  to 50 µl
Delivery information: Supplied without syringe.
Documents
Specifications
  • Description:
    Repeating dispenser, PB600-1