Orders LinkContactUsLinkComponent
Sampling swabs
Sampling swabs
Sampling swabs
Supplier:  DELTALAB
Certificates
Sampling swabs
4 Options Available Below
Product Details & Documents
For safe sampling. Item 115-8250 is longer than conventional swabs and was designed to reach hard-to-reach areas. For example, suitable for use in gynaecology when used with a speculum to remove endocervical cells. Also ideal for veterinary medicine.
  • All swabs have round, tight, tear-resistant heads to ensure perfect sampling without residues
Tip Ø: 5 mm
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Shaft/head
    Ø
    Length
    Sterile
    Availability
    Price
  • 115-8231
    Wooden shafts (only)
    Wooden shafts (only)
    Wood
    2,2 ±0,3 mm
    147 ±0,3 mm
    -
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 25,90 €
    25,90 €
    1 * 1.000 ST
       25,90 €
  • Sampling swabs
    Wood/cotton
    2,2 mm
    150 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 5,90 €
    5,90 €
    1 * 100 ST
       5,90 €
  • Sampling swabs
    Wood/cotton
    2,5 mm
    200 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 20,20 €
    20,20 €
    1 * 100 ST
       20,20 €
  • Sampling swabs
    PP/cotton
    2,5 mm
    200 mm
    -
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 7,50 €
    7,50 €
    1 * 100 ST
       7,50 €