Orders LinkContactUsLinkComponent
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
Certificates
Sponge swabs, Polywipe®
Sponge swabs, Polywipe®
2 Options Available Below
Product Details & Documents
Non-inhibitory sponge material allows effective sampling of larger areas for environmental and hygiene monitoring. Available dry or pre-moistened with sterile diluent. Polywipes are supplied in peel pouches for immediate use, or in a 110 ml blue-capped container for transfer to the laboratory.
  • Blue colour for visibility
  • Triple wrapped and radiation sterilised
Delivery information: Supplied with individual sterile glove for handling.
Product Family Options
Product Information
  • Availability
    Price
  • 720-0144
    Swab containers, 110 ml
    Swab containers, 110 ml
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 192,00 €
    192,00 €
    1 * 30 ST
       192,00 €
  • 720-0145
    Swab peel pouches
    Swab peel pouches
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 197,00 €
    197,00 €
    1 * 50 ST
       197,00 €