Orders LinkContactUsLinkComponent
Stoppers, rubber, conical
Stoppers, rubber, conical
Supplier:   KARTELL
Stoppers, rubber, conical
Supplier:  KARTELL
Certificates
Stoppers, rubber, conical
Stoppers, rubber, conical
6 Options Available Below
Product Details & Documents
Natural red rubber with elastic, tight fitting design.
  • Excellent seal and adhesion
  • Good chemical resistance
  • Non toxic
  • Temperature resistant from −25 to +70 °C
Product Family Options
Product Information
  • No. of Holes
    Height
    Ø bottom
    Ø top
    Availability
    Price
  • 0
    24,0 mm
    14,0 mm
    20,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1,10 €
    1,10 €
    1 * 1 ST
       1,10 €
  • 0
    28,0 mm
    19,0 mm
    26,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2,40 €
    2,40 €
    1 * 1 ST
       2,40 €
  • 0
    47,0 mm
    34,0 mm
    45,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,50 €
    9,50 €
    1 * 1 ST
       9,50 €
  • 0
    50,0 mm
    37,0 mm
    48,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,70 €
    11,70 €
    1 * 1 ST
       11,70 €
  • 0
    60,0 mm
    48,0 mm
    60,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,60 €
    20,60 €
    1 * 1 ST
       20,60 €
  • 0
    70,0 mm
    50,0 mm
    65,0 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €