Stoppers, press down, VacuCap®
About this item
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
- Disposable seals are an inexpensive alternative to standard seals
- Double flanged seal for an impermeable, secure fit
- Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
13 Options Available Below
Product Details & Documents
Ideal closures for glass and plastic reagent, sample and culture tubes, LDPE.
- Disposable seals are an inexpensive alternative to standard seals
- Double flanged seal for an impermeable, secure fit
- Colour coded for sample identification
Flexible to protect from aerosols or highly infectious microorganisms. Samples are protected from cross-contamination and the work environment from infection due to leaks. The double flanged seal guarantees a tight and impermeable fit for a wide range of applications, including centrifuging.
Product Family Options
Product Information
- ColourØ bottomAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.217-0101 - Specifications:
More
Specifications
Cat. No.720-0552 - Specifications:
More
Specifications
Cat. No.217-0098 - Specifications:
More
Specifications
Cat. No.217-0099 - Specifications:
More
Specifications
Cat. No.217-0102 - Specifications:
More
Specifications
Cat. No.720-0551 - Specifications:
More
Specifications
Cat. No.720-0554 - Specifications:
More
Specifications
Cat. No.720-0555 - Specifications:
More
Specifications
Cat. No.217-0103 - Specifications:
More
Specifications
Cat. No.217-0104 - Specifications:
More
Specifications
Cat. No.217-0106 - Specifications:
More
Specifications
Cat. No.217-0105 - Specifications:
More
Specifications
Cat. No.720-0553