Orders LinkContactUsLinkComponent
Stopcocks, three way
Stopcocks, three way
Supplier:   Lenz Laborglas
Stopcocks, three way
Supplier:  Lenz Laborglas
Certificates
Stopcocks, three way
Stopcocks, three way
12 Options Available Below
Product Details & Documents
DURAN®, borosilicate glass, with solid glass or PTFE key.
  • Replaceable NS keys with T-bore
  • Screw thread retaining device
  • Leakage rate compliant with DIN 12540-1
DIN 12554
Documents
Product Family Options
With solid glass key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 1,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,50 €
    43,50 €
    1 * 1 ST
       43,50 €
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,50 €
    43,50 €
    1 * 1 ST
       43,50 €
  • 2,5 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 85,00 €
    85,00 €
    1 * 1 ST
       85,00 €
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 58,00 €
    58,00 €
    1 * 1 ST
       58,00 €
  • 4,0 mm
    24,0 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,70 €
    66,70 €
    1 * 1 ST
       66,70 €
  • 6,0 mm
    29,2 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 93,90 €
    93,90 €
    1 * 1 ST
       93,90 €
With PTFE key
Product Information
  • Hole Ø
    Spigot
    Availability
    Price
  • 1,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,80 €
    51,80 €
    1 * 1 ST
       51,80 €
  • 2,5 mm
    14,5 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 66,80 €
    66,80 €
    1 * 1 ST
       66,80 €
  • 2,5 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,20 €
    69,20 €
    1 * 1 ST
       69,20 €
  • 4,0 mm
    18,8 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 68,80 €
    68,80 €
    1 * 1 ST
       68,80 €
  • 4,0 mm
    24,0 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 107,00 €
    107,00 €
    1 * 1 ST
       107,00 €
  • 6,0 mm
    29,2 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 220,00 €
    220,00 €
    1 * 1 ST
       220,00 €