Orders LinkContactUsLinkComponent
Stopcocks, straight bore
Stopcocks, straight bore
Supplier:   SciLabware
Stopcocks, straight bore
Supplier:  SciLabware
Certificates
Stopcocks, straight bore
5 Options Available Below
Product Details & Documents
PYREX®, borosilicate glass with PTFE key.
  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
Product Family Options
Single bore
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 1 mm
    8 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 124,00 €
    124,00 €
    1 * 5 ST
       124,00 €
  • 2 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 126,00 €
    126,00 €
    1 * 5 ST
       126,00 €
  • 4 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 151,00 €
    151,00 €
    1 * 5 ST
       151,00 €
  • 6 mm
    12 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 240,00 €
    240,00 €
    1 * 5 ST
       240,00 €
Double bore
Product Information
  • Hole Ø
    For tube I-Ø
    Availability
    Price
  • 2,5 mm
    9 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 424,00 €
    424,00 €
    1 * 5 ST
       424,00 €
Accessories
Product Information
  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST102K
    1,5 mm
    Log in for availability
    1 * 5 ST - 95,50 €
Product Information
  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST103K
    2,5 mm
    Log in for availability
    1 * 5 ST - 112,00 €
Product Information
  • Catalog #
    Hole Ø
    Availability
    Price
  • Spare PTFE keys for stopcocks, PYREX®, straight bore
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    BIBBSST104K
    4,0 mm
    Log in for availability
    1 * 5 ST - 159,00 €