Magnetic stirring bars, tapered
: COWIE
PTFE covered, strong Alnico V magnetic core. Smooth surface. Stirring action is similar to oval or pivot ring type stirring bars.
- Excellent chemical resistance
- Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Frequently Bought Together