Orders LinkContactUsLinkComponent
Splash head, vertical, Quickfit®
Splash head, vertical, Quickfit®
19/26 NS, 24/29 NS
Catalog # 201-8691
Supplier:   SciLabware
Splash head, vertical, Quickfit®
Catalog # 201-8691
Supplier:  SciLabware
Splash head, vertical, Quickfit®
Splash head, vertical, Quickfit®
19/26 NS, 24/29 NS
Price
78,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
Usually inserted between a flask and still head to prevent 'carry-over' of raw liquid.
Specifications
  • Cone size:
    24/29 NS
  • Socket size:
    19/26 NS