Order Entry
Belgium
ContactUsLinkComponent
Dry heat indicator tapes
Dry heat indicator tapes
  489-1410
 :  KARTELL
undefined
Dry heat indicator tapes
  489-1410
 :  KARTELL

 

  • Width:
    19 mm
  • Description:
    Adhesive tapes for dry heat sterilisation
  • Length:
    50 m
  • Pk:
    1 Roll

 

 

Adhesive tape.

Specially designed for dry heat sterilisation (30 minutes at 155 °C). Colour change from white to brown.