Orders LinkContactUsLinkComponent
Laboratory marker
Marker, Black
Catalog # 631-0014
Supplier:   MARIENFELD
Laboratory marker
Catalog # 631-0014
Supplier:  MARIENFELD
Laboratory marker
Laboratory marker
Marker, Black
Price
3,90 €
Log in for availability
1 * 1 ST
Product Details & Documents
Permanent marker for indelibly labelling microscope slides and embedding cassettes with a high contrast black ink. Ultra-fine tip. These laboratory markers do not dry out and have a long lifetime.
  • Water resistant, odour-free and non toxic
  • Resistant against most laboratory chemicals
  • No pretreatment of surfaces necessary
Specifications
  • Colour:
    Black
  • Description:
    Marker