Marker pens, sterile, Klerpack™
About this item
Efficient and cost effective option that can save a significant amount of time. Blue or black marker pens are available to meet QA requirements for photocopied documents. They feature alcohol resistant labelling and autoseal ink.
- Polyethylene packaging is easier to wipe, enabling more effective spray and wipe
- Reduces the risk of any initial bioburden
- Gamma irradiated at no less than 25 kGy
Packaging: Marker pens are individually double bagged and packed in a third bag.
0 Options Available Below
Product Details & Documents
Efficient and cost effective option that can save a significant amount of time. Blue or black marker pens are available to meet QA requirements for photocopied documents. They feature alcohol resistant labelling and autoseal ink.
- Polyethylene packaging is easier to wipe, enabling more effective spray and wipe
- Reduces the risk of any initial bioburden
- Gamma irradiated at no less than 25 kGy
Packaging: Marker pens are individually double bagged and packed in a third bag.
Product Family Options