TexWrite® Spiral Cleanroom Notebooks
New Product
Supplier: Texwipe
Jump to:
About this item
TexWrite® cleanroom spiral-bound notebook, college ruled. An economical choice for note taking and data recording in critical environments.
- Printed with low sodium, IPA resistant inks
- Lie flat and pages rotate freely for ease of use
Format: large (279×216 mm), medium (216×140 mm), small (127×76 mm).
: Bag-Within-A-Bag®.
3 Options Available Below
Product Details & Documents
TexWrite® cleanroom spiral-bound notebook, college ruled. An economical choice for note taking and data recording in critical environments.
- Printed with low sodium, IPA resistant inks
- Lie flat and pages rotate freely for ease of use
Format: large (279×216 mm), medium (216×140 mm), small (127×76 mm).
: Bag-Within-A-Bag®.
Product Family Options
Product Information
- ColourModelTypePaper sizeNo. of pagesAvailabilityPrice
- TEXWTX5742Notebook, spiralSpecifications:New ProductNew Product
- TEXWTX5741Notebook, spiralSpecifications:New ProductNew Product
- 818-0129Notebook, spiralSpecifications:New ProductNew Product