Orders LinkContactUsLinkComponent
Sample spoons
Sample spoons
Supplier:   BURKLE
Sample spoons
Supplier:  BURKLE
Certificates
Sample spoons
7 Options Available Below
Product Details & Documents
Made of AISI 304 (1.4301) stainless steel. This specially shaped spoon reaches almost every corner in containers and prevents samples from trickling out.
  • For withdrawing small sample quantities
  • Largely chemical resistant (not against acids and alkalis)
Product Family Options
Product Information
  • Length
    Availability
    Price
  • 300-0019
    Sample spoon
    Sample spoon
    150 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,60 €
    9,60 €
    1 * 1 ST
       9,60 €
  • BURK5320-0180
    Sample spoon
    Sample spoon
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10,30 €
    10,30 €
    1 * 1 ST
       10,30 €
  • 300-0020
    Sample spoon
    Sample spoon
    210 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 11,10 €
    11,10 €
    1 * 1 ST
       11,10 €
  • BURK5320-0250
    Sample spoon
    Sample spoon
    250 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 13,50 €
    13,50 €
    1 * 1 ST
       13,50 €
    Specifications:
    New Product
    New Product
  • BURK5320-0300
    Sample spoon
    Sample spoon
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,20 €
    18,20 €
    1 * 1 ST
       18,20 €
  • 300-0021
    Sample spoon
    Sample spoon
    400 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,60 €
    23,60 €
    1 * 1 ST
       23,60 €
  • BURK5320-0500
    Sample spoon
    Sample spoon
    500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,30 €
    27,30 €
    1 * 1 ST
       27,30 €