Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Safety Glasses
Spectacle cords
Spectacle cords
Spectacle cords
111-8052
:
UVEX
Spectacle cords
111-8052
:
UVEX
Colour:
Black
Description:
Cord for safety spectacles
Seq:
01
Pk:
1
Spectacle cords for use with most safety spectacles.