Safety spectacles, uvex super G
: UVEX
These ultra-light, high tech polymer safety glasses have soft and flexible arms. The Super G ensures a comfortable and pressure-free fit that prevents fatigue when in use.
- Extremely thin and highly break resistant lenses
- High flexibility and high projectile resistance
- Aerodynamic shape provides maximum prevention of fog build-up
- Smooth material transition at the ends of the arms
Weight: 18 g
EN 166-168, EN 170; (EN 172: Anti-UV 5-2,5); 1F
: EN 166-168, EN 170; (EN 172: Anti-UV 5-2.5); 1F
Frequently Bought Together