Orders LinkContactUsLinkComponent
Micro powder spatula set
Micro powder spatula set
Set of 4 micro powder spatulas, 3/4/5/6 mm
Catalog # 231-0196
Supplier:   RSG
Micro powder spatula set
Catalog # 231-0196
Supplier:  RSG
Micro powder spatula set
Micro powder spatula set
Set of 4 micro powder spatulas, 3/4/5/6 mm
Price
26,80 €
Log in for availability
1 * 4 ST
Product Details & Documents
Stainless steel 18/10 blade with black plastic handle.
  • Four-piece set, each with a 3, 4, 5 and 6 mm wide powder spatula
  • Blade has excellent chemical and thermal resistance
  • Blades are semicircular and angled
Specifications
  • Description:
    Set of 4 micro powder spatulas
  • Blade width:
    3/4/5/6 mm