Orders LinkContactUsLinkComponent
Section lifters
Section lifters
Supplier:   RSG
Section lifters
Supplier:  RSG
Certificates
Section lifters
3 Options Available Below
Product Details & Documents
Blade made of stainless steel, wooden handle.
  • Blade with excellent heat resistance
  • Good chemical resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Blade length
    Blade width
    Availability
    Price
  • 150 mm
    50 mm
    20 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 52,90 €
    52,90 €
    1 * 5 ST
       52,90 €
  • 150 mm
    50 mm
    10 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 50,70 €
    50,70 €
    1 * 5 ST
       50,70 €
  • 150 mm
    50 mm
    15 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 51,90 €
    51,90 €
    1 * 5 ST
       51,90 €