Orders LinkContactUsLinkComponent
Spoon spatulas, broad shape
Supplier:   NICKEL ELECTRO
Spoon spatulas, broad shape
Supplier:  NICKEL ELECTRO
Certificates
Spoon spatulas, broad shape
Spoon spatulas, broad shape
4 Options Available Below
Product Details & Documents
Pure nickel (99,6%).
  • One end is flat and spatula-shaped, the other end is spoon-shaped
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Product Family Options
Product Information
  • Length
    Spoon size
    Capacity
    Availability
    Price
  • 120 mm
    20 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,90 €
    19,90 €
    1 * 1 ST
       19,90 €
  • 150 mm
    20 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • 180 mm
    20 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,10 €
    32,10 €
    1 * 1 ST
       32,10 €
  • 210 mm
    20 mm
    2 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,70 €
    40,70 €
    1 * 1 ST
       40,70 €