Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
Spoon spatulas, micro
Spoon spatulas, micro
Stainless steel.
  • One end flattened, the other end spoon-formed
  • Good chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Product Family Options
  • Length
    Spoon size
    Capacity
  • 150 mm
    4×6×1,4 mm
    0,1 ml
    1 * 10 ST
    1 * 10 ST - 139,00 €
    139,00 €
    1 * 10 ST
       139,00 €
  • 6×8×1,5 mm
    0,2 ml
    1 * 10 ST
    1 * 10 ST - 147,00 €
    147,00 €
    1 * 10 ST
       147,00 €