Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Spatulas
Spoon spatula, Nuffield type
Spoon spatula, Nuffield type
Spoon spatula, Nuffield type
231-0170
:
NICKEL ELECTRO
Spoon spatula, Nuffield type
231-0170
:
NICKEL ELECTRO
Length:
140 mm
Pk:
10
Stainless steel. Straight with box joint.
Excellent thermal resistance, autoclavable
Excellent chemical resistance
Quick and easy to clean