Orders LinkContactUsLinkComponent
Double spatulas, micro
 :   RSG
Double spatulas, micro
 :  RSG
Double spatulas, micro
Double spatulas, micro
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Product Family Options
  • Length
    Blade width
  • 150 mm
    3 mm
    1 * 5 ST
    1 * 5 ST - 36,90 €
    36,90 €
    1 * 5 ST
       36,90 €
  • 150 mm
    4 mm
    1 * 1 ST
    1 * 1 ST - 7,40 €
    7,40 €
    1 * 1 ST
       7,40 €
  • 130 mm
    6 mm
    1 * 5 ST
    1 * 5 ST - 36,90 €
    36,90 €
    1 * 5 ST
       36,90 €
  • 150 mm
    6 mm
    1 * 5 ST
    1 * 5 ST - 37,20 €
    37,20 €
    1 * 5 ST
       37,20 €
  • 185 mm
    6 mm
    1 * 5 ST
    1 * 5 ST - 37,90 €
    37,90 €
    1 * 5 ST
       37,90 €
  • 210 mm
    6 mm
    1 * 5 ST
    1 * 5 ST - 38,40 €
    38,40 €
    1 * 5 ST
       38,40 €
  • 130 mm
    4 mm
    1 * 5 ST
    1 * 5 ST - 36,70 €
    36,70 €
    1 * 5 ST
       36,70 €
  • 210 mm
    3 mm
    1 * 5 ST
    1 * 5 ST - 38,00 €
    38,00 €
    1 * 5 ST
       38,00 €
  • 150 mm
    7 mm
    1 * 5 ST
    1 * 5 ST - 37,50 €
    37,50 €
    1 * 5 ST
       37,50 €
  • 185 mm
    4 mm
    1 * 5 ST
    1 * 5 ST - 37,50 €
    37,50 €
    1 * 5 ST
       37,50 €
  • 150 mm
    5 mm
    1 * 5 ST
    1 * 5 ST - 37,00 €
    37,00 €
    1 * 5 ST
       37,00 €