Microarray slides, Gene Frame® II
Supplier: Thermo Fisher Scientific
Certificates
About this item
Microarray slides for in situ hybridisation and amplification with standard microscope slides.
- Gas tight sealing prevents reagent loss due to evaporation
- Withstand temperatures of up to 97 °C
- Frame and coverslip can be removed simultaneously leaving no residue
- Can be autoclaved or sterilised by UV irradiation
Delivery information: All packs are supplied with 100 coverslips.
Frequently Bought Together