Orders LinkContactUsLinkComponent
Cell strainers, Falcon®
Cell strainers, Falcon®
Supplier:   Corning
Cell strainers, Falcon®
Supplier:  Corning
Certificates
Cell strainers, Falcon®
Cell strainers, Falcon®
3 Options Available Below
Product Details & Documents
An easy, ready-to-use way to consistently obtain a more uniform single-cell suspension. Made of a strong nylon mesh and available with 40, 70 or 100 µm pores that are evenly spaced for optimal performance in a variety of applications. A faster and easier alternative to gauze filtration in procedures involving dissociation of cells from either clumps or primary tissue.
  • Moulded, colour-coded PP frame with tab enables easy handling
  • Fits perfectly into a 50 ml Falcon® conical tube or other similarly sized tube
  • Sterilised by gamma irradiation and conveniently available in individual, peel-open packaging
  • Extended lip on strainer enables aseptic handling with forceps
Ordering information: Packaged individually in bags.
Product Family Options
Product Information
  • Availability
    Price
  • 734-0002
    Cell strainers, 40 µm pore size, blue frame
    Cell strainers, 40 µm pore size, blue frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 111,73 €
    111,73 €
    1 * 50 ST
       111,73 €
  • 734-0003
    Cell strainers, 70 µm pore size, white frame
    Cell strainers, 70 µm pore size, white frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 111,73 €
    111,73 €
    1 * 50 ST
       111,73 €
  • 734-0004
    Cell strainers, 100 µm pore size, yellow frame
    Cell strainers, 100 µm pore size, yellow frame
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 111,73 €
    111,73 €
    1 * 50 ST
       111,73 €